Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFP64 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590771
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590771 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590771 Supplier Invitrogen™ Supplier No. PA590771
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MNASSEGESF AGSVQIPGGT TVLVELTPDI HICGICKQQF NNLDAFVAHK QSGCQLTGTS AAAPSTVQFV SEETVPATQT QTTTRTITSE TQTITVSAPE FVFEHGYQTY LPTESNENQT ATVISLPAKS RTKKPTTPPA QKRLNCCYPG CQFKTAYGMK DMERHLKIHT; Positive Samples: 293T, NIH/3T3, Raji; Cellular Location: Nucleus.

ZFP64 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 9 C2H2-type zinc fingers. ZFP64 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFP64
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ZFP64
Gene Accession No. Q99KE8, Q9NTW7
Gene Alias ZFP64; Zfp-64; ZFP64 zinc finger protein; zinc finger protein 338; Zinc finger protein 64; zinc finger protein 64 homolog; ZNF338
Gene Symbols ZFP64
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-170 of human ZFP64 (NP_0606672).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22722, 55734
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.