Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFP64 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111467
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111467 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111467 Supplier Invitrogen™ Supplier No. PA5111467
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SEGRVKIIVG HQVPQANTIV QAAAAAVNIV PPALVAQNPE ELPGNSRLQI LRQVSLIAPP QSSRCPSEAG AMTQPAVLLT THEQTDGA Highest antigen sequence indentity to the following orthologs: Rat - 84%, Mouse - 85%.

ZFP64 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 9 C2H2-type zinc fingers. ZFP64 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFP64
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZFP64
Gene Accession No. Q9NTW7
Gene Alias ZFP64; Zfp-64; ZFP64 zinc finger protein; zinc finger protein 338; Zinc finger protein 64; zinc finger protein 64 homolog; ZNF338
Gene Symbols ZFP64
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human ZFP64. Recombinant protein control fragment (Product #RP-103327).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55734
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.