Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFP69 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569735
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569735 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569735 Supplier Invitrogen™ Supplier No. PA569735
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.

Putative transcription factor that appears to regulate lipid metabolism. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFP69
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZFP69
Gene Accession No. A2A761
Gene Alias Gm1029; Krab2; rCG30900-like; RP11-656D10.2; Zfp63; Zfp63p; Zfp69; ZFP69 zinc finger protein; ZFP69 zinc finger protein A; ZFP69A; Zfp965; zinc finger protein 642; Zinc finger protein 69; Zinc finger protein 69 homolog; zinc finger protein 69 homolog; zinc finger protein ZFP69; zinc finger protein 965; zinc finger protein ZFP69; ZKSCAN23A; ZNF642; ZSCAN54A
Gene Symbols ZFP69
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of mouse ZFP69.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 381549
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.