Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFP69 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111406
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111406 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111406 Supplier Invitrogen™ Supplier No. PA5111406
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MRDVRQAILT HKKRVQETNK FGENIIVHSN VIIEQRHHKY DTPTKRNTYK LDLINHPTSY IRTKTYECNI CE Highest antigen sequence indentity to the following orthologs: Mouse - 53%, Rat - 49%.

Putative transcription factor that appears to regulate lipid metabolism. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFP69
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZFP69
Gene Accession No. Q49AA0
Gene Alias Gm1029; Krab2; rCG30900-like; RP11-656D10.2; Zfp63; Zfp63p; Zfp69; ZFP69 zinc finger protein; ZFP69 zinc finger protein A; ZFP69A; Zfp965; zinc finger protein 642; Zinc finger protein 69; Zinc finger protein 69 homolog; zinc finger protein 69 homolog; zinc finger protein ZFP69; zinc finger protein 965; zinc finger protein ZFP69; ZKSCAN23A; ZNF642; ZSCAN54A
Gene Symbols ZFP69
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human ZFP69. Recombinant protein control fragment (Product #RP-106584).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339559
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.