Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFP82 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563862
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563862 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563862 Supplier Invitrogen™ Supplier No. PA563862
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NHGLKGLILK NDWESTGKIE GQERPQEGYF SSVKMPSEKV SSYQKRTSVT PHQRLH Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 50%.

ZFP82 is a protein coding gene, and may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFP82
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZFP82
Gene Accession No. Q8N141
Gene Alias A030010D05; Kiaa1948; Krab16; Mszf3; ZFP82; Zfp-82; ZFP82 zinc finger protein; zinc finger protein 545; Zinc finger protein 82; zinc finger protein 82 homolog; zinc finger protein 85; ZNF545
Gene Symbols ZFP82
Host Species Rabbit
Immunogen Recombinant protein fragment of Human ZFP82 (Product #RP-100460)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284406
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.