Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZFP82 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569038
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 93%.
ZFP82 is a protein coding gene, and may be involved in transcriptional regulation.
Specifications
ZFP82 | |
Polyclonal | |
Unconjugated | |
ZFP82 | |
A030010D05; Kiaa1948; Krab16; Mszf3; ZFP82; Zfp-82; ZFP82 zinc finger protein; zinc finger protein 545; Zinc finger protein 82; zinc finger protein 82 homolog; zinc finger protein 85; ZNF545 | |
Rabbit | |
Affinity chromatography | |
RUO | |
284406 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N141 | |
ZFP82 | |
synthetic peptide directed towards the C terminal region of human ZFP82. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction