Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFP90 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen ZFP90
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP179411
SDP
View Documents
Novus Biologicals
NBP179411
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

ZFP90 Polyclonal specifically detects ZFP90 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ZFP90
Polyclonal
Rabbit
NP_597715
146198
Synthetic peptide directed towards the middle region of human ZFP90The immunogen for this antibody is ZFP90. Peptide sequence SSLVQHQRIHTGEKPYRCNLCGRSFRHGTSLTQHEVTHSGEKPFQCKECG.
Primary
Western Blot
Unconjugated
RUO
KIAA1954, NK10, zfp-90, zinc finger protein 476, zinc finger protein 756, zinc finger protein 90 homolog, zinc finger protein 90 homolog (mouse), ZNF756
ZFP90
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.