Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFP90 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568548
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568548 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568548 Supplier Invitrogen™ Supplier No. PA568548
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 85%; Rabbit: 100%; Rat: 100%.

ZFP90 may function as a repressor or silencer protein, and most likely exerts its repressing activity upon zinc-dependent binding to DNA. It may be involved in proper spermatogenesis by repressing the expression of genes unnecessary or incompatible with the maintenance of a haploid cell state.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFP90
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZFP90
Gene Accession No. Q61967
Gene Alias 6430515L01Rik; FIK; FOXP3-interacting KRAB domain-containing protein; KIAA1954; KRAB17; mKIAA1954; NK10; Nk10 expressed protein; Zfp64; Zfp83; Zfp90; zfp-90; ZFP90 zinc finger protein; zinc finger protein 476; zinc finger protein 756; zinc finger protein 83; zinc finger protein 90; Zinc finger protein 90 homolog; zinc finger protein NK10; ZNF756
Gene Symbols ZFP90
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human ZFP90 (aa 49-98).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22751
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.