Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZFP90 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | ZFP90 |
---|---|
Concentration | 0.44 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-5221
![]() ![]() |
Proteintech Group Inc
261201AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
ZFP90 may function as a repressor or silencer protein, and most likely exerts its repressing activity upon zinc-dependent binding to DNA. It may be involved in proper spermatogenesis by repressing the expression of genes unnecessary or incompatible with the maintece of a haploid cell state.Specifications
ZFP90 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q4V8A8, Q61967, Q8TF47 | |
146198, 22751, 498945 | |
ZFP90 Fusion Protein Ag23381 | |
Primary | |
-20°C | |
ZFP90 |
0.44 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
KIAA1954, NK10, Zfp 90, ZFP90, Zinc finger protein 90 homolog, ZNF756 | |
Zfp90 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title