Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZFP91 Antibody (8C3D5) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261847
Description
ZFP91 Monoclonal specifically detects ZFP91 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ZFP91 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100- 1:500, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
FLJ57065, ZFP-91, zinc finger protein 91 homolog (mouse), zinc finger protein homologous to Zfp91 in mouse | |
Mouse | |
44.5 kDa | |
0.1 mL | |
80829 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
8C3D5 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
ZFP91 | |
Purified recombinant fragment of human ZFP91 (AA: 162-304) expressed in E. Coli. | |
Protein G purified | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction