Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZFP91 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554960
Description
Immunogen sequence: EEHQSPGGIS SEEEEEEEEE MLISEEEIPF KDDPRDETYK PHLERETPKP RRKSGKVKEE KEKKEIKVEV EVEVKEEENE IREDEE Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 97%.
ZFP91 is a member of the zinc finger family of proteins. The gene product contains C2H2 type domains, which are the classical zinc finger domains found in numerous nucleic acid-binding proteins. In addition to the monocistronic transcript originating from this locus, a co-transcribed variant composed of ZFP91 and CNTF sequence has been identified. The monocistronic and co-transcribed variants encode distinct isoforms. The co-transcription of ZFP91 and CNTF has also been observed in mouse.
Specifications
ZFP91 | |
Polyclonal | |
Unconjugated | |
ZFP91 | |
9130014I08Rik; A530054C17Rik; AL024263; AW545902; cocaine attenuated zinc-finger protein; DMS-8; drug resistance-associated sequence in melanoma 8; DSM-8; E3 ubiquitin-protein ligase ZFP91; FKSG11; Penta Zf protein; penta zinc finger protein; PZF; RING-type E3 ubiquitin transferase ZFP91; Zfp91; Zfp-91; ZFP91 zinc finger protein; ZFP91 zinc finger protein, atypical E3 ubiquitin ligase; zinc finger protein 757; zinc finger protein 91; zinc finger protein 91 homolog; zinc finger protein 91-like protein; zinc finger protein homologous to Zfp91 in mouse; Zinc finger protein PZF; ZNF757 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80829 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96JP5 | |
ZFP91 | |
Recombinant protein corresponding to Human ZFP91. Recombinant protein control fragment (Product #RP-91697). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction