Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZFP92 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP24757325UL
Description
ZFP92 Polyclonal specifically detects ZFP92 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ZFP92 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
zfp-92, zinc finger protein 92 homolog, zinc finger protein 92 homolog (mouse), zinc finger protein homologous to Zfp92 in mouse, ZNF897 | |
Rabbit | |
Affinity Purified | |
RUO | |
139735 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
ZFP92 | |
This antibody was developed against a recombinant protein corresponding to amino acids: YSHLVSLGFSFSKPHLISQLERGEGPWVADIPRTWATAGLHIGDRTQSKTSTSTQKHSGRQLPGADPQGGKEGQAARSSVLQRGAQGLGQSSAAGPQGPKGAEKRYLCQQCGKAFSRSSNLIKHR | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction