Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZFP92 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551801
Description
Immunogen sequence: YSHLVSLGFS FSKPHLISQL ERGEGPWVAD IPRTWATAGL HIGDRTQSKT STSTQKHSGR QLPGADPQGG KEGQAARSSV LQRGAQGLGQ SSAAGPQGPK GAEKRYLCQQ CGKAFSRSSN LIKHR Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 50%.
May be involved in transcriptional regulation.
Specifications
| ZFP92 | |
| Polyclonal | |
| Unconjugated | |
| ZFP92 | |
| ZFP92; Zfp-92; ZFP92 zinc finger protein; zinc finger protein 92 homolog; zinc finger protein homologous to Zfp92 in mouse; ZNF897 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 139735 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A6NM28 | |
| ZFP92 | |
| Recombinant protein corresponding to Human ZFP92. Recombinant protein control fragment (Product #RP-92309). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction