Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFP95 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551857
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551857 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551857 Supplier Invitrogen™ Supplier No. PA551857
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EHHPESGEEA VAVIENIQRE LEERRQQIVA CPDVLPRKMA TPGAVQESCS PHPLTVDTQP EQAPQKPRLL EENALPVLQV PSLPLKDSQE LTASLLSTGS QKLVKIEEVA DVAVSFILEE WGHLDQSQKS LYRDDRKENY GSITS Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 77%.

Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. A member of the Krü;ppel C2H2-type zinc-finger protein family, ZKSCAN5, also known as ZFP95, is a 839 amino acid protein containing thirteen C2H2-type zinc fingers, one KRAB A domain and one SCAN box domain. ZKSCAN5 is located in the nucleus and is highly expressed in testis with lower levels of expression in liver, kidney, brain, striated muscle, lung and heart. Many KRAB A box-containing proteins have been shown to function as transcription repressors, suggesting a possible function of ZKSCAN5. As a result of alternative splicing, three transcripts of ZKSCAN5 exist, however one of those transcripts is specific to testis. The mouse homolog of ZKSCAN5 is differentially expressed in spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFP95
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZKSCAN5
Gene Accession No. Q9Y2L8
Gene Alias AI132486; AI326970; hKraba1; KIAA1015; ZFP95; ZFP-95; Zinc finger protein 95; zinc finger protein 95 homolog; zinc finger protein homologous to Zfp95 in mouse; zinc finger protein with KRAB and SCAN domains 5; zinc finger with KRAB and SCAN domains 5; Zkscan5; ZNF914; ZSCAN37
Gene Symbols ZKSCAN5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZFP95. Recombinant protein control fragment (Product #RP-99734).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23660
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.