Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFPL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540868
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540868 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540868 Supplier Invitrogen™ Supplier No. PA540868
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: NGPIFPPTNL AGPVASALRE KLATVNWARA GLGLPLIDEV VSPEPEPLNT Sequence homology: Cow: 100%; Dog: 92%; Goat: 100%; Guinea Pig: 100%; Horse: 92%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

ZFPL1 is expressed strongly in the exocrine pancreas as a 1.4-kb polyadenylated RNA encoding a putative protein of 310 amino acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFPL1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZFPL1
Gene Accession No. O95159
Gene Alias 1500015B20Rik; D11S750; MCG4; Unknown (protein for MGC:134051); ZFPL1; zinc finger like protein 1; zinc finger protein like 1; zinc finger protein MCG4; Zinc finger protein-like 1; zinc-finger protein in MEN1 region
Gene Symbols ZFPL1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ZFPL1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7542
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.