Learn More
Invitrogen™ ZFPM2 Polyclonal Antibody

Description
Recommended positive controls: HeLa, mouse brain, rat brain. Predicted reactivity: Mouse (95%), Rat (94%), Dog (95%), Pig (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | ZFPM2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.96 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | ZFPM2 |
| Gene Accession No. | Q8CCH7, Q8WW38 |
| Gene Alias | B330005D23Rik; DIH3; Fog2; FOG-2; Friend of GATA 2; friend of GATA protein 2; friend of GATA-1 2; Friend of GATA2; friend of GATA-2; hFOG-2; mFOG-2; SRXY9; transcription factor GATA4, modulator of; ZC2HC11B; Zfpm2; Zinc finger protein 89B; Zinc finger protein multitype 2; Zinc finger protein ZFPM2; zinc finger protein, FOG family member 2; zinc finger protein, multitype 2; ZNF89B |
| Gene Symbols | ZFPM2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.