Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFR Mouse anti-Human, Unconjugated, Clone: 1G2B9, MilliporeSigma™

Catalog No. MABE18751MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE18751MI 100 μg
MABE18752MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABE18751MI Supplier MilliporeSigma Supplier No. MABE1875100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZFR Monoclonal antibody specifically detects ZFR in Human samples. It is validated for Chromatin Immunoprecipitation (ChIP), Immunohistochemistry (Paraffin), Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFR
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1G2B9
Conjugate Unconjugated
Gene Accession No. NP_057191
Gene Alias Zinc finger RNA-binding protein;hZFR;M-phase phosphoprotein homolog
Gene Symbols ZFR
Host Species Mouse
Immunogen Recombinant fragment corresponding to 372 amino acids from the C-terminal half of human Zinc finger RNA-binding protein (ZFR).
Molecular Weight of Antigen ∼120 kDa observed; 117.01 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 100 μg
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity Clone 1G2B9 is a mouse monoclonal antibody tthat detects human Zinc finger RNA-binding protein..
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.