Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540913
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540913 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540913 Supplier Invitrogen™ Supplier No. PA540913
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA5-40913 detects ZFR in WB, IHC and IF applications. Peptide sequence: RQIHKVLGMD PLPQMSQRFN IHNNRKRRRD SDGVDGFEAE GKKDKKDYDN Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

ZFR is critical for Staufen 2 isoform specific nucleocytoplasmic shuttling in neurons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZFR
Gene Accession No. Q96KR1
Gene Alias C920030H05Rik; hnrpa1; hZFR; M-phase phosphoprotein homolog; SPG71; wu:fb25b08; Zfr; zfr protein; ZFR1; zgc:66235; zgc:66403; zinc finger RNA binding protein; zinc finger RNA-binding protein; zinc finger RNA-binding-like protein
Gene Symbols ZFR
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ZFR.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51663
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.