Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFY Polyclonal Antibody

Catalog No. PIPA567210
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567210 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567210 Supplier Invitrogen™ Supplier No. PA567210

Rabbit Polyclonal Antibody

Immunogen sequence: KTHIKTKHSKE MPFKCDICLL TFSDTKEVQQ HTLVHQESKT HQCLHCD Highest antigen sequence identity to the following orthologs - mouse 96%, rat 94%.

This gene encodes a zinc finger-containing protein that may function as a transcription factor. This gene was once a candidate gene for the testis-determining factor (TDF) and was erroneously referred to as TDF.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFY
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZFY
Gene Accession No. P08048
Gene Alias ZFY; zinc finger protein, Y-linked; zinc finger Y-chromosomal protein; ZNF911
Gene Symbols ZFY
Host Species Rabbit
Immunogen Recombinant Human ZFY. Recombinant protein control fragment (Product #RP-105660).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7544
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.