Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZFY1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568796
Description
This target displays homology in the following species: Mouse: 100%.
Probable transcriptional activator. Binds to the consensus sequence 5'-AGGCCY-3'.
Specifications
ZFY1 | |
Polyclonal | |
Unconjugated | |
Zfy1 | |
Y-linked zinc finger protein 1; Zfy1; Zfy-1; zinc finger protein 1, Y linked; zinc finger Y-chromosomal protein 1 | |
Rabbit | |
Protein A | |
RUO | |
22767 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P10925 | |
Zfy1 | |
synthetic peptide directed towards the middle region of mouse ZFY1. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction