Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFYVE1 Monoclonal Antibody (OTI4H7)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA527255

Catalog No. PIMA527255


Only null left
Add to Cart

Description

Description

ZFYVE1 Monoclonal Antibody for Western Blot

The FYVE domain mediates the recruitment of proteins involved in membrane trafficking and cell signaling to phosphatidylinositol 3-phosphate (PtdIns(3)P)-containing membranes. This gene encodes a protein which contains two zinc-binding FYVE domains in tandem. This protein displays a predominantly Golgi, endoplasmic reticulum and vesicular distribution. Alternatively spliced transcript variants have been found for this gene, and they encode two isoforms with different sizes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZFYVE1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9HBF4
ZFYVE1
Human recombinant protein fragment corresponding to amino acids 1-308 of ZFYVE1 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2a
Western Blot
OTI4H7
Unconjugated
ZFYVE1
DFCP1; Double FYVE-containing protein 1; G630053K23; KIAA1589; mKIAA1589; phosphoinositide-binding protein SR3; PP10436; PPP1R172; protein phosphatase 1, regulatory subunit 172; SR3; TAFF1; Tandem FYVE fingers-1; tandem FYVE fingers-1 protein; Zfyve1; Zinc finger FYVE domain-containing protein 1; zinc finger FYVE-type containing 1; zinc finger protein, subfamily 2A, member 1; zinc finger, FYVE domain containing 1; ZNFN2A1
Mouse
Affinity Chromatography
RUO
53349
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.