Learn More
ZFYVE1 Mouse anti-Human, Clone: OTI4B3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810111

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The FYVE domain mediates the recruitment of proteins involved in membrane trafficking and cell signaling to phosphatidylinositol 3-phosphate (PtdIns(3)P)-containing membranes. This gene encodes a protein which contains two zinc-binding FYVE domains in tandem. This protein displays a predomitly Golgi, endoplasmic reticulum and vesicular distribution. Alternatively spliced transcript variants have been found for this gene, and they encode two isoforms with different sizes.Specifications
ZFYVE1 | |
Monoclonal | |
Unconjugated | |
ZFYVE1 | |
DFCP1, PPP1R172, SR3, TAFF1, ZNFN2A1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
53349 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4B3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9HBF4 | |
ZFYVE1 | |
Human recombit protein fragment corresponding to amino acids 1-308 of human ZFYVE1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.