Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFYVE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530638
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530638 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530638 Supplier Invitrogen™ Supplier No. PA530638
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, IMR32, U87-MG. Predicted reactivity: Mouse (96%), Rat (96%), Xenopus laevis (83%), Chicken (92%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The FYVE domain mediates the recruitment of proteins involved in membrane trafficking and cell signaling to phosphatidylinositol 3-phosphate (PtdIns(3)P)-containing membranes. This gene encodes a protein which contains two zinc-binding FYVE domains in tandem. This protein displays a predominantly Golgi, endoplasmic reticulum and vesicular distribution. Alternatively spliced transcript variants have been found for this gene, and they encode two isoforms with different sizes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFYVE1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene ZFYVE1
Gene Accession No. Q9HBF4
Gene Alias DFCP1; Double FYVE-containing protein 1; G630053K23; KIAA1589; mKIAA1589; phosphoinositide-binding protein SR3; PP10436; PPP1R172; protein phosphatase 1, regulatory subunit 172; SR3; TAFF1; Tandem FYVE fingers-1; tandem FYVE fingers-1 protein; Zfyve1; Zinc finger FYVE domain-containing protein 1; zinc finger FYVE-type containing 1; zinc finger protein, subfamily 2A, member 1; zinc finger, FYVE domain containing 1; ZNFN2A1
Gene Symbols ZFYVE1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ZFYVE1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53349
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.