Learn More
Invitrogen™ ZFYVE1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596710
Description
Immunogen sequence: AKYAWSGYVI ECPNCGVVYR SRQYWFGNQD PVDTVVRTEI VHVWPGTDGF LKDNNNAAQR LLDGMNFMAQ SVSELSLGPT KAVTSWLTDQ IAPAYWRPNS QILSCNKCAT SFKDNDTKHH CRACGEGFCD SCSSKTRPVP ERGWGPAPVR VCDNCYEARN VQLAVTEAQV DDEGGTLIAR KVGEAVQNTL GAVVTAIDIP LGLVKDAARP AYWVPDHEIL HCHNCRKEFS IKLSKH Positive Samples: 293T; Cellular Location: Endoplasmic reticulum, Golgi apparatus, Golgi stack.
The FYVE domain mediates the recruitment of proteins involved in membrane trafficking and cell signaling to phosphatidylinositol 3-phosphate (PtdIns(3)P)-containing membranes. This gene encodes a protein which contains two zinc-binding FYVE domains in tandem. This protein displays a predominantly Golgi, endoplasmic reticulum and vesicular distribution. Alternatively spliced transcript variants have been found for this gene, and they encode two isoforms with different sizes.
Specifications
ZFYVE1 | |
Polyclonal | |
Unconjugated | |
ZFYVE1 | |
DFCP1; Double FYVE-containing protein 1; G630053K23; KIAA1589; mKIAA1589; phosphoinositide-binding protein SR3; PP10436; PPP1R172; protein phosphatase 1, regulatory subunit 172; SR3; TAFF1; Tandem FYVE fingers-1; tandem FYVE fingers-1 protein; Zfyve1; Zinc finger FYVE domain-containing protein 1; zinc finger FYVE-type containing 1; zinc finger protein, subfamily 2A, member 1; zinc finger, FYVE domain containing 1; ZNFN2A1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
53349 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
Q9HBF4 | |
ZFYVE1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 500-735 of human ZFYVE1 (NP_0670831). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.