Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZFYVE1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5107317
Description
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
The FYVE domain mediates the recruitment of proteins involved in membrane trafficking and cell signaling to phosphatidylinositol 3-phosphate (PtdIns(3)P)-containing membranes. This gene encodes a protein which contains two zinc-binding FYVE domains in tandem. This protein displays a predominantly Golgi, endoplasmic reticulum and vesicular distribution. Alternatively spliced transcript variants have been found for this gene, and they encode two isoforms with different sizes.
Specifications
ZFYVE1 | |
Polyclonal | |
Unconjugated | |
ZFYVE1 | |
DFCP1; Double FYVE-containing protein 1; G630053K23; KIAA1589; mKIAA1589; phosphoinositide-binding protein SR3; PP10436; PPP1R172; protein phosphatase 1, regulatory subunit 172; SR3; TAFF1; Tandem FYVE fingers-1; tandem FYVE fingers-1 protein; Zfyve1; Zinc finger FYVE domain-containing protein 1; zinc finger FYVE-type containing 1; zinc finger protein, subfamily 2A, member 1; zinc finger, FYVE domain containing 1; ZNFN2A1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
53349 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
Q9HBF4 | |
ZFYVE1 | |
Synthetic peptide directed towards the C-terminal region of human ZFYVE1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction