Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFYVE16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100192
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100192 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100192 Supplier Invitrogen™ Supplier No. PA5100192
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ZFYVE16.

The ZFYVE16 gene encodes endofin, an endosomal protein implicated in regulating membrane trafficking. It is characterized by the presence of a phosphatidylinositol 3-phosphate-binding FYVE domain positioned in the middle of the molecule.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFYVE16
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ZFYVE16
Gene Accession No. Q7Z3T8, Q80U44
Gene Alias AI035632; B130024H06Rik; B130031L15; Endofin; endosomal-associated FYVE domain protein; Endosome-associated FYVE domain protein; endosome-associated FYVE-domain protein; Kiaa0305; mKIAA0305; PPP1R69; protein phosphatase 1, regulatory subunit 69; RGD1564784; ZFYVE16; Zinc finger FYVE domain-containing protein 16; zinc finger FYVE-type containing 16; zinc finger, FYVE domain containing 16
Gene Symbols ZFYVE16
Host Species Rabbit
Immunogen A synthesized peptide derived from human ZFYVE16(Accession Q7Z3T8), corresponding to amino acid residues Q1497-F1539.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 218441, 499508, 9765
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.