Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFYVE16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA525044
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA525044 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA525044 Supplier Invitrogen™ Supplier No. PA525044
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZFYVE16 Polyclonal Antibody for Western Blot, IHC (P), Flow

The ZFYVE16 gene encodes endofin, an endosomal protein implicated in regulating membrane trafficking. It is characterized by the presence of a phosphatidylinositol 3-phosphate-binding FYVE domain positioned in the middle of the molecule.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFYVE16
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ZFYVE16
Gene Accession No. Q7Z3T8
Gene Alias AI035632; B130024H06Rik; B130031L15; Endofin; endosomal-associated FYVE domain protein; Endosome-associated FYVE domain protein; endosome-associated FYVE-domain protein; Kiaa0305; mKIAA0305; PPP1R69; protein phosphatase 1, regulatory subunit 69; RGD1564784; ZFYVE16; Zinc finger FYVE domain-containing protein 16; zinc finger FYVE-type containing 16; zinc finger, FYVE domain containing 16
Gene Symbols ZFYVE16
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 375-404 amino acids from the N-terminal region of human ZFYVE16.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9765
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.