Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFYVE19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530856
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530856 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530856 Supplier Invitrogen™ Supplier No. PA530856
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: K562. Predicted reactivity: Human (99%), Mouse (82%), Rat (84%), Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Key regulator of abscission step in cytokinesis: part of the cytokinesis checkpoint, a process required to delay abscission to prevent both premature resolution of intercellular chromosome bridges and accumulation of DNA damage. Together with CHMP4C, required to retain abscission-competent VPS4 (VPS4A and/or VPS4B) at the midbody ring until abscission checkpoint signaling is terminated at late cytokinesis. Deactivation of AURKB results in dephosphorylation of CHMP4C followed by its dissociation from ZFYVE19/ANCHR and VPS4 and subsequent abscission.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFYVE19
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene ZFYVE19
Gene Accession No. Q96K21
Gene Alias 1500041L05Rik; abscission/NoCut checkpoint regulator; ANCHR; MLL partner containing FYVE domain; MPFYVE; Zfyve19; Zinc finger FYVE domain-containing protein 19; zinc finger FYVE-type containing 19; zinc finger, FYVE domain containing 19
Gene Symbols ZFYVE19
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ZFYVE19. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84936
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.