Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZFYVE21 Antibody (OTI2B12), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274924AF405
Description
ZFYVE21 Monoclonal specifically detects ZFYVE21 in Human samples. It is validated for Western Blot.Specifications
ZFYVE21 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ZFYVE21 | |
Full length recombinant protein of human ZFYVE21 (NP_076976) produced in HEK293T cells. | |
0.1 mL | |
79038.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI2B12 | |
Western Blot | |
FYVE domain containing 21, ZF21MGC2550, zinc finger FYVE domain-containing protein 21, zinc finger, FYVE domain containing 21 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction