Learn More
ZFYVE21 Mouse anti-Human, Clone: OTI4A2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505622

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ZF21 was initially identified as protein that could bind to the cytoplasmic tail of MT1-MMP (Membrane-type 1 matrix metalloproteinase), a potent invasion-promoting protease. ZF21 is a member of a protein family characterized by the presence of a phosphatidylinositol 3-phosphate-binding FYVE domain and regulates focal adhesions (FAs) and cell movement. Knockdown of ZF21 expression resulted in a delay of FA disassembly following induction of synchronous disassembly of FAs by nocodazole treatment, suggesting that ZF21 is involved in FA disassembly. ZF21 contains a noncanonical pleckstrin homology domain that is a possible therapeutic target to treat metastatic cancer.Specifications
ZFYVE21 | |
Monoclonal | |
Unconjugated | |
ZFYVE21 | |
HCVP7TP1, ZF21 | |
Mouse | |
Affinity Chromatography | |
RUO | |
79038 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4A2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9BQ24 | |
ZFYVE21 | |
Full length human recombit protein of human ZFYVE21 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.