Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFYVE21 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563165
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563165 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563165 Supplier Invitrogen™ Supplier No. PA563165
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SGATFLVTFG NSEKPETMTC RLSNNQRYLF LDGDSHYEIE IVHISTVQIL TEGFPPGGGN ARATGMFLQY TVPG Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 84%.

ZF21 was initially identified as protein that could bind to the cytoplasmic tail of MT1-MMP (Membrane-type 1 matrix metalloproteinase), a potent invasion-promoting protease. ZF21 is a member of a protein family characterized by the presence of a phosphatidylinositol 3-phosphate-binding FYVE domain and regulates focal adhesions (FAs) and cell movement. Knockdown of ZF21 expression resulted in a delay of FA disassembly following induction of synchronous disassembly of FAs by nocodazole treatment, suggesting that ZF21 is involved in FA disassembly. ZF21 contains a noncanonical pleckstrin homology domain that is a possible therapeutic target to treat metastatic cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFYVE21
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZFYVE21
Gene Accession No. Q9BQ24
Gene Alias 1110013H04Rik; AW558780; C85416; FYVE domain containing 21; HCV p7-transactivated protein 1; HCVP7TP1; ZF21; Zfyve21; Zinc finger FYVE domain-containing protein 21; zinc finger FYVE-type containing 21; zinc finger, FYVE domain containing 21
Gene Symbols ZFYVE21
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZFYVE21. Recombinant protein control fragment (Product #RP-98355).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79038
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.