Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFYVE27 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100193
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100193 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100193 Supplier Invitrogen™ Supplier No. PA5100193
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ZFYVE27.

This gene encodes a protein with several transmembrane domains, a Rab11-binding domain and a lipid-binding FYVE finger domain. The encoded protein appears to promote neurite formation. A mutation in this gene has been reported to be associated with hereditary spastic paraplegia, however the pathogenicity of the mutation, which may simply represent a polymorphism, is unclear.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFYVE27
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ZFYVE27
Gene Accession No. Q3TXX3, Q5T4F4, Q6P7B7
Gene Alias 2210011N02Rik; 9530077C24Rik; AI426636; AI593546; AI835681; im:7137347; LOW QUALITY PROTEIN: protrudin; PROTRUDIN; RP11-459F3.2; Spastic paraplegia 33 protein; SPG33; zfyve27; zgc:153156; zinc finger FYVE domain containing 27; zinc finger FYVE domain-containing protein 27; zinc finger FYVE-type containing 27; zinc finger, FYVE domain containing 27; zinc finger, FYVE domain containing 27 isoform c
Gene Symbols ZFYVE27
Host Species Rabbit
Immunogen A synthesized peptide derived from human ZFYVE27(Accession Q5T4F4), corresponding to amino acid residues D317-S367.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118813, 309376, 319740
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.