Learn More
Invitrogen™ ZFYVE27 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590843
Description
Immunogen sequence: VEFFRVVSEY RASLQQRMNP KQEEHAFESP PPPDVGGKDG LMDSTPALTP TEDLTPGSVE EAEEAEPDEE FKDAIEETHL VVLEDDEGAP CPAEDELALQ DNGFLSKNEV LRSKVSRLTE RLRKRYPTNN FGNCTGCSAT FSVLKKRRSC SNCGNSFCSR CCSFKVPKSS MGATAPEAQR ETVFVCASCN QTLSK; Positive Samples: DU145; Cellular Location: Cell membrane, Cell projection, Endoplasmic reticulum membrane, Multi-pass membrane protein, Recycling endosome membrane, growth cone membrane.
This gene encodes a protein with several transmembrane domains, a Rab11-binding domain and a lipid-binding FYVE finger domain. The encoded protein appears to promote neurite formation. A mutation in this gene has been reported to be associated with hereditary spastic paraplegia, however the pathogenicity of the mutation, which may simply represent a polymorphism, is unclear.
Specifications
ZFYVE27 | |
Polyclonal | |
Unconjugated | |
ZFYVE27 | |
2210011N02Rik; 9530077C24Rik; AI426636; AI593546; AI835681; im:7137347; LOW QUALITY PROTEIN: protrudin; PROTRUDIN; RP11-459F3.2; Spastic paraplegia 33 protein; SPG33; zfyve27; zgc:153156; zinc finger FYVE domain containing 27; zinc finger FYVE domain-containing protein 27; zinc finger FYVE-type containing 27; zinc finger, FYVE domain containing 27; zinc finger, FYVE domain containing 27 isoform c | |
Rabbit | |
Affinity chromatography | |
RUO | |
118813, 309376 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.88 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q5T4F4, Q6P7B7 | |
ZFYVE27 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 217-411 of human ZFYVE27 (NP_6531893). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.