Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFYVE28 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563454
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563454 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563454 Supplier Invitrogen™ Supplier No. PA563454
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ARFYYADEEL NQVAAELDSL DGRKDPQRCT LLVSQFRSCQ DNVLNIINQI MDECIPQDRA PRDFCVKFPE EIRHDNLAGQ LWFGAECLAA GSIIMNREL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

Negative regulator of epidermal growth factor receptor (EGFR) signaling. Acts by promoting EGFR degradation in endosomes when not monoubiquitinated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFYVE28
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZFYVE28
Gene Accession No. Q9HCC9
Gene Alias 9630058O20Rik; Gm146; hLst2; KIAA1643; Lateral signaling target protein 2 homolog; LST2; LYST2; mKIAA1643; Zfyve28; Zinc finger FYVE domain-containing protein 28; zinc finger FYVE-type containing 28; zinc finger, FYVE domain containing 28
Gene Symbols ZFYVE28
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZFYVE28. Recombinant protein control fragment (Product #RP-104293).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57732
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.