Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZGLP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562064
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562064 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562064 Supplier Invitrogen™ Supplier No. PA562064
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLDRDSKDTQ TRISQKGRRL QPPGTPSAPP QRRPRKQLNP CRGTERVDPG FEGVTLKFQI KPDSSLQIIP TYSLPCSSRS QE Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 50%.

Glucagon is initially synthesized as proglucagon, which is 180 amino acids in length and is cleaved proteolytically. It contains a region corresponding to Glucagon itself, in addition to a signal peptide, spacer regions and two Glucagon-like peptide sequences. GLP1 is encoded by the fourth exon of the six in the human Glucagon gene. It is 37 amino acids in length and has 48% amino acid sequence homology to Glucagon, and is thought to have arisen by gene duplication. The sequence of GLP1 is conserved across many species, however, indicating that it has a particular function. GLP1 is a potent insulin secretagogue, renders pancreatic beta cells 'glucose-competent' and may be useful in the treatment of diabetes mellitus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZGLP1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZGLP1
Gene Accession No. P0C6A0
Gene Alias GATA like protein 1; GATA like protein-1; GATA zinc finger domain containing 3; GATAD3; GATA-like 1; GATA-like protein 1; GATA-type zinc finger protein 1; Glp1; GLP-1; ZGLP1; zinc finger, GATA-like protein 1
Gene Symbols ZGLP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZGLP1. Recombinant protein control fragment (Product #RP-99247).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100125288
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.