Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZGPAT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113756
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113756 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113756 Supplier Invitrogen™ Supplier No. PA5113756
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLREAVVEGD GILPPLRTEA TESDSDSDGT GDSSYARVVG SDAVDSAQSS For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.

Transcription repressor that specifically binds the 5'-GGAG[GA]A[GA]A-3' consensus sequence. Represses transcription by recruiting the chromatin multiprotein complex NuRD to target promoters. Negatively regulates expression of EGFR, a gene involved in cell proliferation, survival and migration. Its ability to repress genes of the EGFR pathway suggest it may act as a tumor suppressor. Able to suppress breast carcinogenesis. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZGPAT
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZGPAT
Gene Accession No. Q8N5A5
Gene Alias 1500006I01Rik; BC021513; G patch domain-containing protein 6; GPATC6; GPATCH6; KIAA1847; RGD1310801; ZC3H9; ZC3HDC9; ZGPAT; Zinc finger and G patch domain-containing protein; zinc finger CCCH domain-containing protein 9; zinc finger CCCH-type and G-patch domain containing; zinc finger CCCH-type with G patch domain; zinc finger CCCH-type with G patch domain-containing protein; zinc finger, CCCH-type with G patch domain; zinc finger, CCCH-type with G patch domain isoform c; zinc finger, CCCH-type with G-patch domain; ZIP
Gene Symbols ZGPAT
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ZGPAT.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84619
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.