Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZHX1 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB600244AF532
Description
ZHX1 Polyclonal antibody specifically detects ZHX1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
ZHX1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
zona pellucida B protein, zona pellucida glycoprotein 4ZBP, Zona pellucida protein B, zona pellucida sperm-binding protein 4, Zp-4, ZPBZP1 | |
The immunogen this antibody was made to, maps to a region between residues 1-50 of human zinc fingers and homeoboxes 1 using the TrEMBL entry Q9UKY1 (GeneID 11244). | |
0.1 mL | |
Apoptosis, Cancer, Transcription Factors and Regulators, Tumor Suppressors | |
11244 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction