Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZHX1 Monoclonal Antibody (5E5)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524439
Description
Peptide Sequence: SSSMNGLSSL RKRGRGRPKG RGRGRPRGRP RGSKRINNWD RGPSLIKFKT GTAILKDYYL KRKFLNEQDL DELVNKSHMG YEQVREWFAE RQRRSELGI.
The members of the zinc fingers and homeoboxes gene family are nuclear homodimeric transcriptional repressors that interact with the A subunit of nuclear factor-Y (NF-YA) and contain two C2H2-type zinc fingers and five homeobox DNA-binding domains. This gene encodes member 1 of this gene family. In addition to forming homodimers, this protein heterodimerizes with members 2 and 3 of the zinc fingers and homeoboxes family. Alternative splicing results in multiple transcript variants encoding the same protein.
Specifications
ZHX1 | |
Monoclonal | |
Unconjugated | |
ZHX1 | |
Zhx1; zinc finger and homeodomain protein 1; zinc fingers and homeobox 1; zinc fingers and homeoboxes 1; zinc fingers and homeoboxes protein 1; zinc-fingers and homeoboxes 1 | |
Mouse | |
Protein A | |
RUO | |
11244 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA | |
5E5 | |
PBS with no preservative; pH 7.4 | |
Q9UKY1 | |
ZHX1 | |
ZHX1 (NP_009153, 731 a.a. ~ 829 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction