Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZHX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501729080 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9080 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9080 Supplier Proteintech Group Inc Supplier No. 139031AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The members of the zinc fingers and homeoboxes gene family are nuclear homodimeric transcriptional repressors that interact with the A subunit of nuclear factor-Y (NF-YA) and contain two C2H2-type zinc fingers and five homeobox DNA-binding domains. This gene encodes member 1 of this gene family. In addition to forming homodimers, this protein heterodimerizes with members 2 and 3 of the zinc fingers and homeoboxes family. Alternative splicing results in multiple transcript variants encoding the same protein.

Specifications

Antigen ZHX1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZHX1
Gene Accession No. P70121, Q8R515, Q9UKY1
Gene Alias ZHX1, zinc fingers and homeoboxes 1
Gene Symbols Zhx1
Host Species Rabbit
Immunogen ZHX1 Fusion Protein Ag4874
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11244, 171159, 22770
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.