Learn More
ZHX2 Mouse anti-Human, Clone: OTI9A5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810688

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The members of the zinc fingers and homeoboxes gene family are nuclear homodimeric transcriptional repressors that interact with the A subunit of nuclear factor-Y (NF-YA) and contain two C2H2-type zinc fingers and five homeobox DNA-binding domains. This gene encodes member 2 of this gene family. In addition to forming homodimers, this protein heterodimerizes with member 1 of the zinc fingers and homeoboxes family.Specifications
ZHX2 | |
Monoclonal | |
Unconjugated | |
ZHX2 | |
AFR1, RAF | |
Mouse | |
Affinity Chromatography | |
RUO | |
22882 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI9A5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y6X8 | |
ZHX2 | |
Human recombit protein fragment corresponding to amino acids 595-837 of human ZHX2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.