Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZHX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120626
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120626 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120626 Supplier Invitrogen™ Supplier No. PA5120626
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: U-251MG, 293T, Raji, LO2, Mouse brain. Immunogen sequence: SDHRYRCQRG IVHITSESLA KDQLAIAASR HGRTYHAYPD FAPQKFKEKT QGQVKILEDS FLKSSFPTQA ELDRLRVETK LSRREIDSWF SERRKLRDSM EQAVLDSMGS GKKGQDVGAP NGALSRLDQL SGAQLTSSLP SPSPAIAKSQ EQVHLLRSTF ARTQWPTPQE YDQLAAKTGL VRTEIVRWFK ENRCLLKTGT VKWMEQYQHQ PMADDHGYDA VARKATKPMA ESPKNGGDVV PQYYKDPKKL CEEDLEKLVT RVKVGSEPAK DCLPAKPSEA TSDRSEGSSR DGQGSDENEE SSVVDYVEVT VGEEDAISDR SDSWSQAAAE GVSELAESDS DCVPAEAGQA.

Zinc-fingers and homeobox (ZHX) proteins are transcription factors that interact with the activation domain of the A subunit of nuclear factor-Y (NF-YA). ZHX1-3 are ubiquitously expressed proteins expressed in various tissues. They act as transcriptional repressors and localize to the nucleus. The ZHX proteins contain two Cys(2)-His(2)-type zinc-finger motifs and five homeodomains (HDs). These domains allow the ZHX proteins to form homodimers, but they can also form heterodimers with each other. However, this dimerization is not required for repressor activity. Hypermethylation-mediated silencing of ZHX2 is an epigenetic event involved in hepatocellular carcinoma (HCC).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZHX2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ZHX2
Gene Accession No. Q80VX4, Q8C0C0, Q9Y6X8
Gene Alias AFP regulator 1; AFR1; Afr-1; alpha fetoprotein regulation 1, adult; alpha-fetoprotein regulator 1; KIAA0854; LOW QUALITY PROTEIN: zinc fingers and homeoboxes protein 2; mKIAA0854; Raf; Regulator of AFP; transcription factor ZHX2; Zhx2; Zinc finger and homeodomain protein 2; zinc fingers and homeoboxes 2; zinc fingers and homeoboxes protein 2; zinc-fingers and homeoboxes 2
Gene Symbols ZHX2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 488-837 of human ZHX2 (NP_055758.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22882, 314988, 387609
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.