Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZHX3 Antibody (PCRP-ZHX3-1D11), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314225FR
This item is not returnable.
View return policy
Description
ZHX3 Monoclonal antibody specifically detects ZHX3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
ZHX3 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
23051 | |
Store at 4°C in the dark. | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
PCRP-ZHX3-1D11 | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
FLJ93221, Sp38, zona pellucida binding protein, zona pellucida-binding protein 1, ZPBP1zona pellucida binding protein 1 | |
Recombinant full-length human ZHX3 protein (Uniprot: Q9H4I2) | |
0.1 mL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction