Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZHX3 Antibody (PCRP-ZHX3-1G3), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314230MFV450
Description
ZHX3 Monoclonal antibody specifically detects ZHX3 in Human samples. It is validated for Flow CytometrySpecifications
ZHX3 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
23051 | |
Store at 4C in the dark. | |
IgG2a |
Flow Cytometry | |
PCRP-ZHX3-1G3 | |
Flow Cytometry | |
FLJ93221, Sp38, zona pellucida binding protein, zona pellucida-binding protein 1, ZPBP1zona pellucida binding protein 1 | |
Recombinant full-length human ZHX3 protein (Uniprot: Q9H4I2) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction