Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZHX3 Monoclonal Antibody (1D9)
GREENER_CHOICE

Catalog No. PIMA524444
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524444 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524444 Supplier Invitrogen™ Supplier No. MA524444
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: EDLQNLCDKT QMSSQQVKQW FAEKMGEETR AVADTGSEDQ GPGTGELTAV HKGMGDTYSE VSENSESWEP RVPEASSEPF DTSSPQAGRQ LET.

This gene encodes a member of the zinc fingers and homeoboxes (ZHX) gene family. The encoded protein contains two C2H2-type zinc fingers and five homeodomains and forms a dimer with itself or with zinc fingers and homeoboxes family member 1. In the nucleus, the dimerized protein interacts with the A subunit of the ubiquitous transcription factor nuclear factor-Y and may function as a transcriptional repressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZHX3
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1D9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ZHX3
Gene Accession No. Q9H4I2
Gene Alias 1810059C13Rik; 4932418O04Rik; 9530010N21Rik; KIAA0395; mKIAA0395; Tix1; triple homeobox 1; triple homeobox protein 1; Zhx3; Zinc finger and homeodomain protein 3; zinc fingers and homeoboxes 3; zinc fingers and homeoboxes protein 3
Gene Symbols ZHX3
Host Species Mouse
Immunogen ZHX3 (NP_055850.1, 863 a.a. ~ 955 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23051
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.