Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZHX3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565440
Description
Immunogen sequence: MFNTVIQSVPQ PTITVLNTPL VASAGNVQHL IQAALPGHVV GQPEGTGGGL LVTQPLMANG LQATSSPLPL TVTSVPKQPG VAPINTVCSN TTSAVKVVNA AQSLLTACPS ITSQAFLDAS IYKNKKSHEQ LSALKGSFCR NQFPGQ Highest antigen sequence identity to the following orthologs - mouse 92%, rat 93%.
This gene encodes a member of the zinc fingers and homeoboxes (ZHX) gene family. The encoded protein contains two C2H2-type zinc fingers and five homeodomains and forms a dimer with itself or with zinc fingers and homeoboxes family member 1. In the nucleus, the dimerized protein interacts with the A subunit of the ubiquitous transcription factor nuclear factor-Y and may function as a transcriptional repressor.
Specifications
ZHX3 | |
Polyclonal | |
Unconjugated | |
ZHX3 | |
1810059C13Rik; 4932418O04Rik; 9530010N21Rik; KIAA0395; mKIAA0395; Tix1; triple homeobox 1; triple homeobox protein 1; Zhx3; Zinc finger and homeodomain protein 3; zinc fingers and homeoboxes 3; zinc fingers and homeoboxes protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23051 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H4I2 | |
ZHX3 | |
Recombinant Human ZHX3. Recombinant protein control fragment (Product #RP-106118). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction