Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zic-1 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501990817
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0817 100 μg
50-199-0818 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0817 Supplier Rockland Immunochemicals Supplier No. 200401159
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This is an IgG preparation of whole rabbit antiserum purified by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. This antibody is directed against Zic-1 from mouse. In general, this antibody also detects human Zic-1. Cross-reactivity with other species is likely but has not been determined.

This gene encodes a member of the ZIC family of C2H2-type zinc finger proteins. Members of this family are important during development. Aberrant expression of this gene is seen in medulloblastoma, a childhood brain tumor. This gene is closely linked to the gene encoding zinc finger protein of the cerebellum 4, a related family member on chromosome 3. This gene encodes a transcription factor that can bind and transactivate the apolipoprotein E gene.

Specifications

Antigen Zic-1
Applications ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene ZIC1
Gene Accession No. P46684, Q15915
Gene Alias CRS6; odd-paired homolog; ZIC; Zic family member 1; Zic family member 1 (odd-paired homolog, Drosophila); zic protein member 1; ZIC1; Zinc finger protein 201; Zinc finger protein of the cerebellum 1; Zinc finger protein ZIC 1; ZNF201
Gene Symbols ZIC1
Host Species Rabbit
Immunogen The whole rabbit serum used to produce this IgG fraction antibody was prepared by repeated immunizations with an 18 aa synthetic peptide from a region near the N-Terminus of mouse Zic-1. This domain is completely conserved in human ZIC-1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22771, 7545
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.