Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZIC-2Rabbit anti-Canine, Chimpanzee, Human, Mouse, Rat, Polyclonal, Rockland™
SDP

Catalog No. 501991809
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1809 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-1809 Supplier Rockland Immunochemicals Supplier No. 600401848
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against mouse ZIC2 protein. A BLAST analysis was used to suggest reactivity with this protein from human, chimpanzee, mouse, rat and dog based on 100% homology for the immunogen sequence. Expect cross reactivity with ZIC2 from chicken, zebrafish and frog as the core 16 amino acid residues within the immunogen sequence show very high degrees of homology with ZIC2 from these sources. Cross reactivity with ZIC2 homologues from other sources has not been determined.

ZIC2 (zinc finger protein of the cerebellum 2) is a transcriptional repressor and may regulate tissue specific expression of dopamine receptor D1. In mouse, knockout of Zic1 leads to aplasia of cerebellum and skeletal abnormalities. Mutation in Zic2 and Zic3 in mouse and/or human cause holoprosencephaly and heterotaxis, respectively. Holoprosencephaly is the most common structural anomaly of the human brain.

Specifications

Antigen ZIC-2
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene ZIC2
Gene Accession No. O95409, Q62520
Gene Alias BOS_12680; cerebellum zinc finger protein 2; GENA 29; HPE5; Ku; kumba; odd-paired homolog; Zic family member 2; Zic family member 2 (odd-paired homolog, Drosophila); ZIC2; Zinc finger protein of the cerebellum 2; Zinc finger protein ZIC 2
Gene Symbols ZIC2
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22772, 361096, 467311, 485533, 7546
Target Species Canine, Chimpanzee, Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.