Learn More
Invitrogen™ ZIC1 Monoclonal Antibody (1A8)

Description
Peptide Sequence: GHLLFPGLHE QAAGHASPNV VNGQMRLGFS GDMYPRPEQY GQVTSPRSEH YAAPQLHGYG PMNVNMAAHH GAGA.
Specifications
Specifications
| Antigen | ZIC1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1A8 |
| Concentration | 0.35 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ZIC1 |
| Gene Accession No. | Q15915 |
| Gene Alias | CRS6; odd-paired homolog; ZIC; Zic family member 1; Zic family member 1 (odd-paired homolog, Drosophila); zic protein member 1; ZIC1; Zinc finger protein 201; Zinc finger protein of the cerebellum 1; Zinc finger protein ZIC 1; ZNF201 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.