Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZIC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578235
Description
Positive Control: U87-MG, SK-N-SH, IMR32, SK-N-AS Predicted Reactivity: Mouse (100%), Rat (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the ZIC family of C2H2-type zinc finger proteins. Members of this family are important during development. Aberrant expression of this gene is seen in medulloblastoma, a childhood brain tumor. This gene is closely linked to the gene encoding zinc finger protein of the cerebellum 4, a related family member on chromosome 3. This gene encodes a transcription factor that can bind and transactivate the apolipoprotein E gene.
Specifications
ZIC1 | |
Polyclonal | |
Unconjugated | |
ZIC1 | |
CRS6; odd-paired homolog; ZIC; Zic family member 1; Zic family member 1 (odd-paired homolog, Drosophila); zic protein member 1; ZIC1; Zinc finger protein 201; Zinc finger protein of the cerebellum 1; Zinc finger protein ZIC 1; ZNF201 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7545 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2.2 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q15915 | |
ZIC1 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human ZIC1. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction