Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZIC1 Recombinant Rabbit Monoclonal Antibody (6B5O9)
GREENER_CHOICE

Catalog No. PIMA537918
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537918 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537918 Supplier Invitrogen™ Supplier No. MA537918
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: C6, SH-SY5Y Immunogen sequence: MLLDAGPQYP AIGVTTFGAS RHHSAGDVAE RDVGLGINPF ADGMGAFKLN PSSHELASAG QTAFTSQAPG YAAAAALGHH HHPGHVGSYS SAAFNSTRDF.

This gene encodes a member of the ZIC family of C2H2-type zinc finger proteins. Members of this family are important during development. Aberrant expression of this gene is seen in medulloblastoma, a childhood brain tumor. This gene is closely linked to the gene encoding zinc finger protein of the cerebellum 4, a related family member on chromosome 3. This gene encodes a transcription factor that can bind and transactivate the apolipoprotein E gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZIC1
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 6B5O9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZIC1
Gene Accession No. P46684, Q15915
Gene Alias CRS6; odd-paired homolog; ZIC; Zic family member 1; Zic family member 1 (odd-paired homolog, Drosophila); zic protein member 1; ZIC1; Zinc finger protein 201; Zinc finger protein of the cerebellum 1; Zinc finger protein ZIC 1; ZNF201
Gene Symbols ZIC1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ZIC1 (Q15915).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22771, 64618, 7545
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.